Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetic acid, 98%, Thermo Scientific Chemicals
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA138781000
Price (MYR)
449.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Chemical Identifiers
CAS59-48-3
IUPAC Name1H-indol-2-ol
Molecular FormulaC8H7NO
InChI KeyJHFAEUICJHBVHB-UHFFFAOYSA-N
SMILESOC1=CC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to dark cream to cream to brown
Identification (FTIR)Conforms
FormPowder
Melting Point (clear melt)120.0-130.0?C
Assay (GC)≥97.0%