DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
DL-Thioctic Acid, 98+%
Thermo Scientific Chemicals
DL-Thioctic Acid, 98+%
CAS: 1077-28-7 | C8H14O2S2 | 206.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA138720050
Price (MYR)
557.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
557.00
Add to cart
Specifications
Chemical Name or MaterialDL-Thioctic acid
CAS1077-28-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.