Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiocarbanilide, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA138701000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialThiocarbanilide
Name Note98%
CAS102-08-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.