Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
CAS: 1600-44-8 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA138440250
Price (MYR)
717.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
717.00
Add to cart
Chemical Identifiers
CAS4618-18-2
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyJCQLYHFGKNRPGE-FCVZTGTOSA-N
SMILESOC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
Loss on drying=<2 % (1 g, 105°C, 3 h)