1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Tetramethylbenzene, 97+%, Thermo Scientific Chemicals
CAS: 95-93-2 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA138370050
Price (MYR)
192.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
192.00
Add to cart
Chemical Identifiers
CAS505-48-6
IUPAC Nameoctanedioic acid
Molecular FormulaC8H14O4
InChI KeyTYFQFVWCELRYAO-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point141°C to 144°C
Appearance (Color)White to cream
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %