Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
Tetrahydrofurfuryl alcohol, 98%, Thermo Scientific Chemicals
CAS: 97-99-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass bottle
Catalog number FSA138180025
Price (MYR)
579.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
579.00
Add to cart
Chemical Identifiers
CAS57-48-7
IUPAC Name(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
Molecular FormulaC6H12O6
InChI KeyBJHIKXHVCXFQLS-UYFOZJQFSA-N
SMILESOC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Water Content (Karl Fischer Titration)<0.5% (non-U.S. specification)
Optical Rotation-89.0° ± 3.0° (c=2 in water, non-U.S. specification)
Assay (unspecified)≥98.5 to ≤101.5%
Appearance (Color)White