Tetracosane, 99%, Thermo Scientific Chemicals
Tetracosane, 99%, Thermo Scientific Chemicals
Tetracosane, 99%, Thermo Scientific Chemicals
Tetracosane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetracosane, 99%, Thermo Scientific Chemicals
CAS: 646-31-1 | C24H50 | 338.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA138041000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Residue after evaporation=<0.002 %
Appearance (Form)Liquid