DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals
CAS: 133-37-9 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA137870025
Price (MYR)
2,157.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,157.00
Add to cart
Chemical Identifiers
CAS626-67-5
IUPAC Namehydrogen 1-methylpiperidine chloride
Molecular FormulaC6H14ClN
InChI KeyQDUXDCXILAPLAG-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)>98.5%
Refractive Index1.4360-1.4390 @ 20?C