L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Tartaric acid, 99+%, Thermo Scientific Chemicals
CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number FSA137850025
Price (MYR)
881.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
881.00
Add to cart
Specifications
Chemical Name or MaterialL(+)-Tartaric acid
CAS87-69-4
Melting Point168.0°C to 172.0°C
Health Hazard 1Danger
ColorWhite or Colorless