2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-chloroquinoxaline, 97%, Thermo Scientific Chemicals
CAS: 1448-87-9 | C8H5ClN2 | 164.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA136190050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS57-09-0
IUPAC Namehexadecyltrimethylazanium bromide
Molecular FormulaC19H42BrN
InChI KeyLZZYPRNAOMGNLH-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO4>=99.0 %
Appearance (Color)White