3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3'-Chloroacetophenone, 98%, Thermo Scientific Chemicals
CAS: 99-02-5 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA134840250
Price (MYR)
636.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
636.00
Add to cart
Chemical Identifiers
CAS23100-12-1
IUPAC Name6-chloropyridine-3-carbaldehyde
Molecular FormulaC6H4ClNO
InChI KeyAFWWKZCPPRPDQK-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%