Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
View all
Plastic drum
Catalog number FSA134340010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red-brown to brown to black
Assay1.00 to 1.20 (I/Cl ratio of the Wijs solution
Insoluble matter=<0.005 %
Appearance (Form)Liquid or solid
Assayprepared from the reagent)
Filter aid
RUO – Research Use Only