(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
(±)-erythro-Aleuritic acid, 95%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
FSA13373025025 g
Catalog number FSA133730250
Price (MYR)
884.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
884.00
Add to cart
Chemical Identifiers
CAS40691-33-6
IUPAC Namedichloropalladiumbis(ylium); bis(tris(2-methylphenyl)phosphane)
Molecular FormulaC42H42Cl2P2Pd
InChI KeyGMLMZLGOMMIIMA-UHFFFAOYSA-L
SMILESCl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Elemental Analysis>=94 % (ex C)
Appearance (Color)Yellow to orange