Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA133610011
Price (MYR)
309.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
309.00
Add to cart
Chemical Identifiers
CAS107-75-5
IUPAC Name7-hydroxy-3,7-dimethyloctanal
Molecular FormulaC10H20O2
InChI KeyWPFVBOQKRVRMJB-UHFFFAOYNA-N
SMILESCC(CCCC(C)(C)O)CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Density(20°C) 0.916 to 0.924 g/ml
Appearance (Color)Clear colorless
Refractive index1.4460 to 1.4520 (20°C, 589 nm)
Appearance (Form)Liquid