5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA132860025
Price (MYR)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS5965-83-3, 97-05-2
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
CAS5965-83-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only