5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA132860010
Price (MYR)
750.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
750.00
Add to cart
Chemical Identifiers
CAS1822-51-1
IUPAC Namehydrogen 4-(chloromethyl)pyridine chloride
Molecular FormulaC6H7Cl2N
InChI KeyZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)Pale cream to cream to yellow to pale brown
Melting Point (clear melt)163.0-170.0?C
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Indicator
RUO – Research Use Only