beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
beta-Sitosterol, with ca. 10% campesterol, ca. 75% beta-sitosterol, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number FSA132721000
Price (MYR)
513.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
513.00
Add to cart
Chemical Identifiers
CAS18641-71-9
IUPAC Name2,4-dimethylheptan-3-one
Molecular FormulaC9H18O
InChI KeyMMEXTUAHJZOQKN-UHFFFAOYNA-N
SMILESCCCC(C)C(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4135-1.4165 @ 20?C