Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
CAS: 533-31-3 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA132690250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10217-52-4
IUPAC Namehydrazine
Molecular FormulaH4N2
InChI KeyOAKJQQAXSVQMHS-UHFFFAOYSA-N
SMILESNN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration with HCl63.6 to 64.6 % (N2H4)
Density(20°C) 1.030 to 1.033 g/ml
Refractive index1.4285 to 1.4315 (20°C, 589 nm)
Titration with HCl99.4 to 100.9 % (N2H4.H2O)