Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sesamol, 98%, Thermo Scientific Chemicals
CAS: 533-31-3 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA132690050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSesamol
CAS533-31-3
Melting Point63.0°C to 65.0°C
ColorWhite to Beige
Recommended StorageDarkens on storage particularly when stored in the light and at elevated temperatures