Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals
CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA132641000
Price (MYR)
246.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
246.00
Add to cart
Chemical Identifiers
CAS10022-68-1
IUPAC Namecadmium(2+) bis(nitric acid)
Molecular FormulaCdH2N2O6
InChI KeyIZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric>=99.0 %
Copper (Cu)=<20 ppm
Lead (Pb)=<50 ppm
Solubility(10 % in water) Clear colorless