Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
50 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA131780025
Price (MYR)
1,314.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,314.00
Add to cart
Chemical Identifiers
CAS867-55-0
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO498.5 to 101.5 %
Appearance (Form)Crystalline powder
pH6 to 9 (1% solution)
Solubility(10% in water) Clear colorless