Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
50 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA131780010
Price (MYR)
633.00
Add to cart
Quantity:
1 L
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
633.00
Add to cart
Chemical Identifiers
CAS109-59-1
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4085-1.4115 @ 20?C