Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrazole, 98%, pure, Thermo Scientific Chemicals
CAS: 288-13-1 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA131740250
Price (MYR)
394.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Chemical Identifiers
CAS100-69-6
IUPAC Name2-ethenylpyridine
Molecular FormulaC7H7N
InChI KeyKGIGUEBEKRSTEW-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5490 to 1.5510 (20°C, 589 nm)
Stabilizer0.1 % tert.-butylcatechol