Propionic anhydride, 99, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals
Propionic anhydride, 99, Thermo Scientific Chemicals
CAS: 123-62-6 | C6;H1;0;O3; | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number FSA131522500
Price (MYR)
260.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
260.00
Add to cart
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSiO₂:Al₂O₃ mole ratio 5.1:1
CommentSurface area 900 m²/g