Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propargylamine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA131460050
Price (MYR)
681.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
681.00
Add to cart
Chemical Identifiers
CAS2739-98-2
IUPAC Nameethyl 2-(pyridin-2-yl)acetate
Molecular FormulaC9H11NO2
InChI KeyIUDKTVXSXWAKJO-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear pale yellow to yellow
Refractive Index1.4955-1.4995 @ 20?C