1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Catalog number FSA131421000
Price (MYR)
1,071.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,071.00
Add to cart
Chemical Identifiers
CAS7365-45-9
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular FormulaC8H18N2O4S
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILESOCCN1CCN(CCS(O)(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. specification)
Assay from Supplier's CofA≥98.5% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Appearance (Color)White