1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Catalog number FSA131420250
Price (MYR)
455.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
455.00
Add to cart
Specifications
Chemical Name or Material1,3-Propane sultone
CAS1120-71-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause cancer.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.