1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Propane sultone, 97%, Thermo Scientific Chemicals
CAS: 1120-71-4 | C3H6O3S | 122.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
100 g
Catalog number FSA131420010
Price (MYR)
9,239.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
9,239.00
Add to cart
Chemical Identifiers
CAS63231-67-4
IUPAC Namedihydroxysilanone
Molecular FormulaH2O3Si
InChI KeyIJKVHSBPTUYDLN-UHFFFAOYSA-N
SMILESO[Si](O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Loss on Drying≤2.0%
Particle SizePore diameter (av.): 130-170 angstroms
Particle SizeReport particle size distribution (microns, typical 40-75)