(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
CAS: 7785-70-8 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA131261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS86-93-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%