Pimelic Acid, 98%, Thermo Scientific Chemicals
Pimelic Acid, 98%, Thermo Scientific Chemicals
Pimelic Acid, 98%, Thermo Scientific Chemicals
Pimelic Acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pimelic Acid, 98%, Thermo Scientific Chemicals
CAS: 111-16-0 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA131231000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4938-52-7
IUPAC Namehept-1-en-3-ol
Molecular FormulaC7H14O
InChI KeyPZKFYTOLVRCMOA-UHFFFAOYNA-N
SMILESCCCCC(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4315-1.4355 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%