Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA131072500
Price (MYR)
521.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
521.00
Add to cart
Chemical Identifiers
CAS7646-78-8
IUPAC Nametin(IV) chloride
Molecular FormulaCl4H4Sn
InChI KeyHTGSJGUTBKFNJC-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
AppearanceMay develop some turbidity or precipitate
Titration Argentometric98.5 to 101.5 %
Free chlorine=<10 ppm
Arsenic (As)=<40 ppm