Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA131070025
Price (MYR)
1,343.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,343.00
Add to cart
Chemical Identifiers
CAS50-91-9
IUPAC Name5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H11FN2O5
InChI KeyODKNJVUHOIMIIZ-RRKCRQDMSA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point145°C to 153°C
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=99.0 %