Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalic acid, 99%, Thermo Scientific Chemicals
CAS: 88-99-3 | C8H6O4 | 166.132 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA131070010
Price (MYR)
705.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
705.00
Add to cart
Specifications
Chemical Name or MaterialPhthalic acid
CAS88-99-3
Melting Point200.0°C to 205.0°C
Health Hazard 1Danger