Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyl ether, 99%, Thermo Scientific Chemicals
CAS: 101-84-8 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA130600010
Price (MYR)
429.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
429.00
Add to cart
Chemical Identifiers
CAS623-50-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4170 to 1.4200 (20°C, 589 nm)