Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA130362500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22739-76-0
IUPAC Name(²H₇)propan-2-(²H)ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-PIODKIDGSA-N
SMILES[2H]OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Water=<0.05 %
Appearance (Color)Clear colorless
D-Enrichment>=99.0 %