Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
Glass bottle
Catalog number FSA130360500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS81012-87-5
IUPAC Namedisodium dihydrate phosphono ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC9H18N3Na2O16P3
InChI KeyKNSYTSUGTLJHPS-ODQFIEKDSA-L
SMILESO.O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentReference Constants: ε₂₆₀ (pH 7, DBR): 7.4 x 10³
Molar extinction coefficient (molar absorptivity)εmax (pH 2.0): [13.0 ? 52] x 10³
Spectral Ratio(pH 2.0): A₂₅₀/A₂₆₀: 0.45 ? 0.05
Assay (UV/vis)(pH 2.0): ≥ 96.0%
CommentReference Constants: εmax (pH 7, NRC): 9.1 x 10³