Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylboronic acid, 98+%, may contain varying amounts of anhydride, Thermo Scientific Chemicals
CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA130360100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5707-04-0
IUPAC Name(phenylselanyl)chlorane
Molecular FormulaC6H5ClSe
InChI KeyWJCXADMLESSGRI-UHFFFAOYSA-N
SMILESCl[Se]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Orange to dark red
Melting point59°C to 64°C (Dec)
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Argentometric>=97.5 %