2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, Thermo Scientific Chemicals
2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
25 L
Packaging:
View all
Glass bottle
Catalog number FSA130240025
Price (MYR)
752.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
752.00
Add to cart
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total trace metal impurities=<0.005 %
Appearance (Form)Powder or crystals