1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA130131000
Price (MYR)
2,883.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,883.00
Add to cart
Chemical Identifiers
CAS170141-63-6
IUPAC Name1-[3-(trifluoromethoxy)phenyl]ethan-1-one
Molecular FormulaC9H7F3O2
InChI KeyUYHTUQHYGKAYJM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(OC(F)(F)F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4500-1.4550 @ 20?C
Indicator
RUO – Research Use Only