1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA130130250
Price (MYR)
1,036.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,036.00
Add to cart
Specifications
Chemical Name or Material1, 10-Phenanthroline monohydrate
CAS5144-89-8
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only