Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenanthrene, 97%, Thermo Scientific Chemicals
CAS: 85-01-8 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA130090050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS546-68-9
IUPAC Nametitanium(4+) tetrakis(propan-2-olate)
Molecular FormulaC12H28O4Ti
InChI KeyVXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Color (APHA)≤25
Assay (unspecified)Ti: 27.50-28.30% (as TiO2)
Chloride content≤100ppm
Specific Gravity0.950-0.960 @ 25?C