Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA129601000
Price (MYR)
223.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
223.00
Add to cart
Chemical Identifiers
CAS3125-64-2
IUPAC Name1-isothiocyanato-3-methoxybenzene
Molecular FormulaC8H7NOS
InChI KeyWHBYCPUKGYEYFU-UHFFFAOYSA-N
SMILESCOC1=CC=CC(=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6370-1.6410 @ 20?C
Assay (GC)>97.5%
Appearance (Color)Clear colorless to yellow