Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
Packaging:
View all
Plastic drum
Catalog number FSA129600050
Price (MYR)
2,707.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,707.00
Add to cart
Chemical Identifiers
CAS33240-34-5
IUPAC Namebromo(cyclopentyl)magnesium
Molecular FormulaC5H9BrMg
InChI KeyPZOJUJCALDFFHC-UHFFFAOYSA-M
SMILESBr[Mg]C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Appearance (Form)Solution
Titration Grignard1.90 to 2.20 M