Octanoyl chloride, 99%, Thermo Scientific Chemicals
Octanoyl chloride, 99%, Thermo Scientific Chemicals
Octanoyl chloride, 99%, Thermo Scientific Chemicals
Octanoyl chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octanoyl chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA129435000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride content0.001%, max
Heavy Metals (as Pb)5ppm, max
Appearance (Color)White to pale cream
Formcrystalline powder
Residue on ignition0.02%, max