DL-2-Octanol, 97%, Thermo Scientific Chemicals
DL-2-Octanol, 97%, Thermo Scientific Chemicals
DL-2-Octanol, 97%, Thermo Scientific Chemicals
DL-2-Octanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-2-Octanol, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA129411000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-90-2
IUPAC NameN-(4-hydroxyphenyl)acetamide
Molecular FormulaC8H9NO2
InChI KeyRZVAJINKPMORJF-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point168°C to 172°C