Octacosane, 99+%, Thermo Scientific Chemicals
Octacosane, 99+%, Thermo Scientific Chemicals
Octacosane, 99+%, Thermo Scientific Chemicals
Octacosane, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octacosane, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA129271000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS61-78-9
IUPAC Name2-[(4-aminophenyl)formamido]acetic acid
Molecular FormulaC9H10N2O3
InChI KeyHSMNQINEKMPTIC-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(=O)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point197°C to 204°C (Dec)
Titration with NaOH98.5 to 101.5 %
Appearance (Color)White to beige to grey to light brown