n-Nonane, 99%, pure, Thermo Scientific Chemicals
n-Nonane, 99%, pure, Thermo Scientific Chemicals
n-Nonane, 99%, pure, Thermo Scientific Chemicals
n-Nonane, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Nonane, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA129115000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS72745-76-7
IUPAC Name2-methyl-1,3-benzoxazol-5-amine
Molecular FormulaC8H8N2O
InChI KeyFECYTFWPNCBIHC-UHFFFAOYSA-N
SMILESCC1=NC2=CC(N)=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
Melting point70°C to 76°C
HPLC>=96.0 %