2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
Thermo Scientific Chemicals
2-Nitrophenylhydrazine, 97%, moistened with ca 30% water
CAS: 3034-19-3 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA12883010010 gGlass bottle
Catalog number FSA128830100
Price (MYR)
560.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
560.00
Add to cart
Chemical Identifiers
CAS7757-82-6
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Chloride (Cl)=<3000 ppm (as NaCl)
Appearance (Color)White
Titration after Ion exchange98.5 to 101.5 %
Water=<0.1 %