4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA128750250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl acetate
Name Note97%
CAS830-03-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger