4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals
CAS: 830-03-5 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA128750050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS823-85-8
IUPAC Namehydrogen (4-fluorophenyl)hydrazine chloride
Molecular FormulaC6H8ClFN2
InChI KeyFEKUXLUOKFSMRO-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Assay (Argentometric Titration)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder